Thiopicolinamide 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Pyridine-2-carbothioamide, 2-Pyridinethioamide, 2-Pyridylthiocarboxamide

OR964116-100G OR964116-25G
APOSOR964116-100GEA 1160 DKK
APOSOR964116-100G APOSOR964116-25G
Thiopicolinamide 97%
Thiopicolinamide
Formel: C₆H₆N₂S
Molekylvægt: 138.19 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00087576
CAS-nummer: 5346-38-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR