4-Methoxy-2-naphthylamine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Methoxy-β-naphthylamine

OR963955-100MG OR963955-1G OR963955-250MG
APOSOR963955-100MGEA 1260 DKK
APOSOR963955-100MG APOSOR963955-1G APOSOR963955-250MG
4-Methoxy-2-naphthylamine 95%
4-Methoxy-2-naphthylamine
Formel: C₁₁H₁₁NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00056592
CAS-nummer: 2764-95-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR