N-(2-Aminoethyl)glycine 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-((2-Aminoethyl)amino)acetic acid, N-(2-Aminoethyl)-Gly-OH, Ethylenediamine-N-monoacetic Acid

OR963284-1G OR963284-250MG OR963284-5G
APOSOR963284-1GEA 513 DKK
APOSOR963284-1G APOSOR963284-250MG APOSOR963284-5G
N-(2-Aminoethyl)glycine 97%
N-(2-Aminoethyl)glycine
Formel: C₄H₁₀N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00144824
CAS-nummer: 24123-14-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR