N-Formyl-L-valine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: For-Val-OH

OR963260-1G OR963260-25G OR963260-5G
APOSOR963260-1GEA 828 DKK
APOSOR963260-1G APOSOR963260-25G APOSOR963260-5G
N-Formyl-L-valine 95%
N-Formyl-L-valine
Formel: C₆H₁₁NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00021029
CAS-nummer: 4289-97-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR