1-Isopropyl-1H-pyrazol-4-amine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR962707-10G OR962707-1G OR962707-5G
APOSOR962707-10GEA 3280 DKK
APOSOR962707-10G APOSOR962707-1G APOSOR962707-5G
1-Isopropyl-1H-pyrazol-4-amine 98%
1-Isopropyl-1H-pyrazol-4-amine
Formel: C₆H₁₁N₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08700622
CAS-nummer: 97421-16-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR