Quinoline-6-carbonitrile 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR959498-1G OR959498-5G
APOSOR959498-1GEA 226 DKK
APOSOR959498-1G APOSOR959498-5G
Quinoline-6-carbonitrile 96%
Quinoline-6-carbonitrile
Formel: C10H6N2
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09999543
CAS-nummer: 23395-72-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR