Quinolin-8-ylacetic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR953534-100MG OR953534-1G OR953534-250MG
APOSOR953534-100MGEA 3170 DKK
APOSOR953534-100MG APOSOR953534-1G APOSOR953534-250MG
Quinolin-8-ylacetic acid 95%
Quinolin-8-ylacetic acid
Formel: C₁₁H₉NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08691265
CAS-nummer: 152150-04-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR