(2-Chlorophenyl)acetamide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(2-Chlorophenyl)acetamide

OR953412-1G OR953412-5G
APOSOR953412-1GEA 436 DKK
APOSOR953412-1G APOSOR953412-5G
(2-Chlorophenyl)acetamide 98%
(2-Chlorophenyl)acetamide
Formel: C₈H₈ClNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00176724
CAS-nummer: 10268-06-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR