4-(2-Thienyl)smørsyre 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-(Thiophen-2-yl)butanoic acid

OR953352-100G OR953352-25G OR953352-5G
APOSOR953352-100GEA 6490 DKK
APOSOR953352-100G APOSOR953352-25G APOSOR953352-5G
4-(2-Thienyl)smørsyre 97%
4-(2-Thienyl)smørsyre
Formel: C₈H₁₀O₂S
Molekylvægt: 170.23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005463
CAS-nummer: 4653-11-6

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR