4-(Chloromethyl)benzamide 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR952720-1G OR952720-5G
APOSOR952720-1GEA 1340 DKK
APOSOR952720-1G APOSOR952720-5G
4-(Chloromethyl)benzamide 96%
4-(Chloromethyl)benzamide
Formel: C₈H₈ClNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00175950
CAS-nummer: 84545-14-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR