Ethyl-4-hydroxypyrimidine-5-carboxylate 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR952315-1G
OR952315-5G
APOSOR952315-1GEA
162
DKK
APOSOR952315-1G
APOSOR952315-5G
Ethyl-4-hydroxypyrimidine-5-carboxylate 97%
Ethyl-4-hydroxypyrimidine-5-carboxylate
Formel:
C₇H₈N₂O₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00235034 CAS-nummer: 4786-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...