4-Phenylcyclohexanamine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR952070-1G OR952070-250MG OR952070-5G
APOSOR952070-1GEA 2110 DKK
APOSOR952070-1G APOSOR952070-250MG APOSOR952070-5G
4-Phenylcyclohexanamine 95%
4-Phenylcyclohexanamine
Formel: C₁₂H₁₇N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00130038
CAS-nummer: 19992-45-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR