Glycine dimethylamide 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Amino-N,N-dimethylacetamide

OR952034-1G OR952034-5G
APOSOR952034-1GEA 647 DKK
APOSOR952034-1G APOSOR952034-5G
Glycine dimethylamide 97%
Glycine dimethylamide
Formel: C₄H₁₀N₂O
Molekylvægt: 102.14 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06655265
CAS-nummer: 1857-19-8
UN nummer: 2735
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR