6-Chloro-1,2,3,4-tetrahydroquinoline 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR951504-1G OR951504-5G
APOSOR951504-1GEA 610 DKK
APOSOR951504-1G APOSOR951504-5G
6-Chloro-1,2,3,4-tetrahydroquinoline 98%
6-Chloro-1,2,3,4-tetrahydroquinoline
Formel: C₉H₁₀ClN
Molekylvægt: 167.6400 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09034966
CAS-nummer: 49716-18-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR