3-(4-Methylpiperidin-1-yl)propylamine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR951471-1G OR951471-5G
APOSOR951471-1GEA 1640 DKK
APOSOR951471-1G APOSOR951471-5G
3-(4-Methylpiperidin-1-yl)propylamine 98%
3-(4-Methylpiperidin-1-yl)propylamine
Formel: C₉H₂₀N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00129008
CAS-nummer: 6241-30-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR