2-Oxo-1,2,3,4-tetrahydroquinoline-6-carboxylic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR951214-1G OR951214-5G
APOSOR951214-1GEA 218 DKK
APOSOR951214-1G APOSOR951214-5G
2-Oxo-1,2,3,4-tetrahydroquinoline-6-carboxylic acid 95%
2-Oxo-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
Formel: C₁₀H₉NO₃
Molekylvægt: 191.1834 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08271842
CAS-nummer: 70639-77-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR