p-Tolyldiethanolamine 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N,N-Bis(2-hydroxyethyl)-p-toluidine, 2,2'-(4-Methylphenylimino)diethanol, 2,2'-(p-Tolylimino)diethanol, 2,2'-(p-Tolylazanediyl)diethanol
OR950575-100G
OR950575-1KG
OR950575-500G
APOSOR950575-100GEA
436
DKK
APOSOR950575-100G
APOSOR950575-1KG
APOSOR950575-500G
p-Tolyldiethanolamine 95%
p-Tolyldiethanolamine
Formel:
C₁₁H₁₇NO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00020580 CAS-nummer: 3077-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...