Methyl acetylsalicylate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR950288-25G OR950288-500G
APOSOR950288-25GEA 269 DKK
APOSOR950288-25G APOSOR950288-500G
Methyl acetylsalicylate 95%
Methyl acetylsalicylate
Formel: C₁₀H₁₀O₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014978
CAS-nummer: 580-02-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR