5-Methyl-2-thenoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-Methylthiophene-2-carboxylic acid

OR9459-100G OR9459-25G OR9459-5G
APOSOR9459-100GEA 822 DKK
APOSOR9459-100G APOSOR9459-25G APOSOR9459-5G
5-Methyl-2-thenoic acid
5-Methyl-2-thenoic acid
Formel: C₆H₆O₂S
Molekylvægt: 142.18 g/mol
Smeltepunkt: 135 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005439
CAS-nummer: 1918-79-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR