5-Chloro-1-methyl-1H-indole-2-carboxylic acid ≥98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR943010-1G OR943010-5G
APOSOR943010-1GEA 665 DKK
APOSOR943010-1G APOSOR943010-5G
5-Chloro-1-methyl-1H-indole-2-carboxylic acid ≥98%
5-Chloro-1-methyl-1H-indole-2-carboxylic acid
Formel: C₁₀H₈ClNO₂
Molekylvægt: 209.63 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03848205
CAS-nummer: 59908-47-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR