N-tert-Butylmethacrylamide 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR939185-25G
OR939185-5G
APOSOR939185-25GEA
1140
DKK
APOSOR939185-25G
APOSOR939185-5G
N-tert-Butylmethacrylamide 95%
N-tert-Butylmethacrylamide
Formel:
C₈H₁₅NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00043639 CAS-nummer: 6554-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...