2-(Propylamino)ethanol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR938537-1G OR938537-5G
APOSOR938537-1GEA 0 DKK
APOSOR938537-1G APOSOR938537-5G
2-(Propylamino)ethanol 95%
2-(Propylamino)ethanol
Formel: C₅H₁₃NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002844
CAS-nummer: 16369-21-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR