3-tert-Butylbenzaldehyde 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR938360-1G OR938360-5G
APOSOR938360-1GEA 194 DKK
APOSOR938360-1G APOSOR938360-5G
3-tert-Butylbenzaldehyde 95%
3-tert-Butylbenzaldehyde
Formel: C₁₁H₁₄O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11100027
CAS-nummer: 23039-28-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR