1-(Ammoniooxy)propane chloride 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR938341-1G OR938341-250MG
APOSOR938341-1GEA 5780 DKK
APOSOR938341-1G APOSOR938341-250MG
1-(Ammoniooxy)propane chloride 95%
1-(Ammoniooxy)propane chloride
Formel: C₃H₁₀ClNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00231456
CAS-nummer: 6084-54-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR