2-Bromo-5-nitrofuran 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR937994-1G
OR937994-250MG
APOSOR937994-1GEA
2350
DKK
APOSOR937994-1G
APOSOR937994-250MG
2-Bromo-5-nitrofuran 95%
2-Bromo-5-nitrofuran
Formel:
C₄H₂BrNO₃ Molekylvægt: 191.97 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00142879 CAS-nummer: 823-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...