Cyclopentylmalonic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR937726-10G
OR937726-1G
OR937726-5G
APOSOR937726-10GEA
5130
DKK
APOSOR937726-10G
APOSOR937726-1G
APOSOR937726-5G
Cyclopentylmalonic acid 95%
Cyclopentylmalonic acid
Formel:
C₈H₁₂O₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00059515 CAS-nummer: 5660-81-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...