1,4-Bis(2-hydroxyisopropyl)benzene 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR937183-1G
OR937183-25G
OR937183-5G
APOSOR937183-1GEA
0
DKK
APOSOR937183-1G
APOSOR937183-25G
APOSOR937183-5G
1,4-Bis(2-hydroxyisopropyl)benzene 95%
1,4-Bis(2-hydroxyisopropyl)benzene
Formel:
C₁₂H₁₈O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009827 CAS-nummer: 2948-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...