3-Methylhippuric acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR936872-25G
OR936872-5G
APOSOR936872-25GEA
872
DKK
APOSOR936872-25G
APOSOR936872-5G
3-Methylhippuric acid 95%
3-Methylhippuric acid
Formel:
C₁₀H₁₁NO₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00044399 CAS-nummer: 27115-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...