Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: TBTA, Tris((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)amine

OR932877-1G OR932877-250MG OR932877-5G
APOSOR932877-1GEA 1180 DKK
APOSOR932877-1G APOSOR932877-250MG APOSOR932877-5G
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Formel: C₃₀H₃₀N₁₀
Molekylvægt: 530.64 g/mol
Smeltepunkt: 132…143 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD09265124
CAS-nummer: 510758-28-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR