5,7-Dichloro-1H-imidazo[4,5-b]pyridine 96%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR931839-1G
OR931839-250MG
APOSOR931839-1GEA
535
DKK
APOSOR931839-1G
APOSOR931839-250MG
5,7-Dichloro-1H-imidazo[4,5-b]pyridine 96%
5,7-Dichloro-1H-imidazo[4,5-b]pyridine
Formel:
C₆H₃Cl₂N₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11111407 CAS-nummer: 24485-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...