O-Phenyl chlorthioformat 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR931661-25G OR931661-5G
APOSOR931661-25GEA 556 DKK
APOSOR931661-25G APOSOR931661-5G
O-Phenyl chlorthioformat 95%
O-Phenyl chlorthioformat
Formel: C₇H₅ClOS
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004920
CAS-nummer: 1005-56-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR