N-Methyl-DL-alanine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Me-DL-Ala-OH, H-N-Me-DL-Ala-OH, N-DL-Me-Ala-OH

OR930477-100G OR930477-1KG OR930477-500G
APOSOR930477-100GEA 933 DKK
APOSOR930477-100G APOSOR930477-1KG APOSOR930477-500G
N-Methyl-DL-alanine
N-Methyl-DL-alanine
Formel: C₄H₉NO₂
Smeltepunkt: 317 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00063136
CAS-nummer: 600-21-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR