Isopropyl 4-hydroxybenzoat 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Isopropylparaben, Isopropyl parahydroxybenzoate, Isopropyl p-Hydroxybenzoate, 4-Hydroxybenzoic acid isopropyl ester, Isopropyl Paraben, Isopropyl 4-Hydroxybenzoate

OR930029-25G OR930029-5G
APOSOR930029-25GEA 2030 DKK
APOSOR930029-25G APOSOR930029-5G
Isopropyl 4-hydroxybenzoat 95%
Isopropyl 4-hydroxybenzoat
Formel: C₁₀H₁₂O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016468
CAS-nummer: 4191-73-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR