Ethyl-N-methyl-β-alaninate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR929512-1G OR929512-25G
APOSOR929512-1GEA 164 DKK
APOSOR929512-1G APOSOR929512-25G
Ethyl-N-methyl-β-alaninate 98%
Ethyl-N-methyl-beta-alaninate
Formel: C₆H₁₃NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00672194
CAS-nummer: 2213-08-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR