7-Methylindol-3yl-eddikesyre 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(7-Methyl-1H-indol-3-yl)acetic acid

OR929183-1G OR929183-250MG
APOSOR929183-1GEA 1820 DKK
APOSOR929183-1G APOSOR929183-250MG
7-Methylindol-3yl-eddikesyre 95%
7-Methylindol-3yl-eddikesyre
Formel: C₁₁H₁₁NO₂
Molekylvægt: 189.21
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00047183
CAS-nummer: 5435-36-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR