1-(1H-Pyrrolo[2,3-b]pyridin-5-yl)ethanone 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR927674-1G OR927674-250MG
APOSOR927674-1GEA 3260 DKK
APOSOR927674-1G APOSOR927674-250MG
1-(1H-Pyrrolo[2,3-b]pyridin-5-yl)ethanone 95%
1-(1H-Pyrrolo[2,3-b]pyridin-5-yl)ethanone
Formel: C₉H₈N₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09266225
CAS-nummer: 944937-14-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR