2-Chloro-N-methylnicotinamide 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR927612-1G OR927612-5G
APOSOR927612-1GEA 414 DKK
APOSOR927612-1G APOSOR927612-5G
2-Chloro-N-methylnicotinamide 97%
2-Chloro-N-methylnicotinamide
Formel: C₇H₇ClN₂O
Molekylvægt: 170.6 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08691934
CAS-nummer: 52943-20-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR