3-(Chloromethyl)-5-cyclopropylisoxazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR927159-1G
OR927159-5G
APOSOR927159-1GEA
763
DKK
APOSOR927159-1G
APOSOR927159-5G
3-(Chloromethyl)-5-cyclopropylisoxazole
3-(Chloromethyl)-5-cyclopropylisoxazole
Formel:
C₇H₈ClNO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11053990 CAS-nummer: 1060817-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...