3-Methyl-2-phenylpyridin 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR926358-1G OR926358-25G OR926358-5G
APOSOR926358-1GEA 0 DKK
APOSOR926358-1G APOSOR926358-25G APOSOR926358-5G
3-Methyl-2-phenylpyridin 95%
3-Methyl-2-phenylpyridine
Formel: C₁₂H₁₁N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006283
CAS-nummer: 10273-90-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR