2-Methoxynaphthalen 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR925439-100G
OR925439-500G
APOSOR925439-100GEA
183
DKK
APOSOR925439-100G
APOSOR925439-500G
2-Methoxynaphthalen 95%
2-Methoxynaphthalen
Formel:
C₁₁H₁₀O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004061 CAS-nummer: 93-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...