1-Piperidinepentanol 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR925020-1G
OR925020-5G
APOSOR925020-1GEA
1530
DKK
APOSOR925020-1G
APOSOR925020-5G
1-Piperidinepentanol 95%
1-Piperidinepentanol
Formel:
C₁₀H₂₁NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03093640 CAS-nummer: 2937-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...