o-Tolyl acetate 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Acetic acid o-Tolyl ester, Acetic acid o-Cresyl ester, o-Cresyl acetate
OR924691-100G
OR924691-500G
APOSOR924691-100GEA
1290
DKK
APOSOR924691-100G
APOSOR924691-500G
o-Tolyl acetate 98%
o-Tolyl acetate
Formel:
C₉H₁₀O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00053704 CAS-nummer: 533-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...