3-Chlorobutyric acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR924352-10G OR924352-5G
APOSOR924352-10GEA 1910 DKK
APOSOR924352-10G APOSOR924352-5G
3-Chlorobutyric acid 95%
3-Chlorobutyric acid
Formel: C₄H₇ClO₂
Molekylvægt: 122.55 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00020496
CAS-nummer: 1951-12-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR