Ethyl(1-methylpiperidin-4-yl)amine 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR923609-1G
OR923609-250MG
APOSOR923609-1GEA
6320
DKK
APOSOR923609-1G
APOSOR923609-250MG
Ethyl(1-methylpiperidin-4-yl)amine 95%
Ethyl(1-methylpiperidin-4-yl)amine
Formel:
C₈H₁₈N₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07801141 CAS-nummer: 876717-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...