4-Ethylamino-1-butanol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-(Ethylamino)butan-1-ol, N-Ethylbutanolamine

OR923539-25G OR923539-5G
APOSOR923539-25GEA 1960 DKK
APOSOR923539-25G APOSOR923539-5G
4-Ethylamino-1-butanol 95%
4-Ethylamino-1-butanol
Formel: C₆H₁₅NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00671534
CAS-nummer: 39216-86-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR