2-(Dimethylamino)ethyl acetate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Acetic Acid 2-(Dimethylamino)ethyl Ester

OR922974-100G OR922974-25G
APOSOR922974-100GEA 3270 DKK
APOSOR922974-100G APOSOR922974-25G
2-(Dimethylamino)ethyl acetate 95%
2-(Dimethylamino)ethyl acetate
Formel: C₆H₁₃NO₂
Molekylvægt: 131.18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041912
CAS-nummer: 1421-89-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR