Allyl 4-Hydroxybenzoate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Hydroxybenzoic Acid Allyl Ester, Allylparaben, Allyl 4-hydroxybenzoate

OR922681-25G OR922681-5G
APOSOR922681-25GEA 915 DKK
APOSOR922681-25G APOSOR922681-5G
Allyl 4-Hydroxybenzoate 95%
Allyl-4-hydroxybenzoate
Formel: C₁₀H₁₀O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08276268
CAS-nummer: 18982-18-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR