Diisopropyl sulphate 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Diisopropyl sulphate
OR921753-10G
OR921753-1G
OR921753-5G
APOSOR921753-10GEA
5350
DKK
APOSOR921753-10G
APOSOR921753-1G
APOSOR921753-5G
Diisopropyl sulphate 95%
Diisopropyl sulphate
Formel:
C₆H₁₄O₄S Molekylvægt: 182.23 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01682898 CAS-nummer: 2973-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...