5-Phenyl-o-toluidine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Methyl-[1,1'-biphenyl]-3-amine, 3-Amino-4-methylbiphenyl

OR921484-1G OR921484-5G
APOSOR921484-1GEA 698 DKK
APOSOR921484-1G APOSOR921484-5G
5-Phenyl-o-toluidine 95%
5-Phenyl-o-toluidine
Formel: C₁₃H₁₃N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00270126
CAS-nummer: 80938-67-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR