2-(4-Chlorobutoxy)tetrahydropyran 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR921289-1G OR921289-25G OR921289-5G
APOSOR921289-1GEA 933 DKK
APOSOR921289-1G APOSOR921289-25G APOSOR921289-5G
2-(4-Chlorobutoxy)tetrahydropyran 95%
2-(4-Chlorobutoxy)tetrahydropyran
Formel: C₉H₁₇ClO₂
Molekylvægt: 192.68 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00142812
CAS-nummer: 41302-05-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR